missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Human , BB700, Clone: OKT3, BD Biosciences™
Mouse Monoclonal Antibody
1905.00 SEK - 3595.00 SEK
Specifications
Antigen | CD3 |
---|---|
Clone | OKT3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Blue 700 (BB700) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17227634
|
BD Biosciences
566819 |
25 Tests |
1905.00 SEK
25 tests |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17217634
|
BD Biosciences
566818 |
100 Tests |
3595.00 SEK
100 tests |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow CytometrySpecifications
CD3 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Sheep Erythrocyte Rosette-purified Human T Cells | |
Primary |
OKT3 | |
Monoclonal | |
Mouse | |
Human | |
CD3E; CD3e; T-cell surface antigen T3/Leu-4 epsilon; T3E; TCRE | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB700 under optimum conditions, and unconjugated antibody and free BD Horizon BB700 were removed. |