Läs mer
CD3 Mouse, anti-Human, R718, Clone: UCHT1, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566953
Beskrivning
The UCHT1 monoclonal antibody specifically binds to the human CD3ε-chain, a 20-kDa subunit of the CD3/T cell antigen receptor complex. CD3ε is expressed on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody is mitogenic for CD3ε-positive cells when used in conjunction with costimulatory agents such as pokeweed mitogen or anti-CD28 antibody. CD3 plays a central role in signal transduction during antigen recognition. The UCHT1 antibody stains both surface and intracellular CD3ε unlike the other CD3 clone, HIT3a, that stains only extracellular CD3ε.
Specifikationer
| CD3 | |
| Monoclonal | |
| R718 | |
| 100 Tests | |
| Primary | |
| Refrigerate Upon Arrival |
| Flow Cytometry | |
| UCHT1 | |
| Mouse | |
| RUO | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.