Learn More
CD300c Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553184
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PWLRDFHDPI VEVEVSVFPA GTTTASSPQS SMGTSGPPTK LPVHTWPSVT RKDSPEPSPH PGSLFSNVR Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 36%.
CD300c Polyclonal antibody specifically detects CD300c in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
CD300c | |
Polyclonal | |
Unconjugated | |
CD300C | |
CD300 antigen-like family member C; CD300c; CD300c antigen; CD300c molecule; Clm6; CLM-6; CMRF35; CMRF-35; CMRF35 antigen; CMRF35 leukocyte immunoglobulin-like receptor; CMRF35A; CMRF-35A; CMRF35A leukocyte immunoglobulin-like receptor; CMRF35A1; CMRF35-A1; CMRF-35-like molecule 6; CMRF35-like molecule 6; Gcom1; IgSF16; immunoglobulin superfamily member 16; LIR; RGD1565161 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10871 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q08708 | |
CD300C | |
Recombinant protein corresponding to Human CD300c | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only