missing translation for 'onlineSavingsMsg'
Learn More

CD300e (IREM-2) Mouse anti-Human, PE, Clone: UP-H2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-3007-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15566956

  • 1442.00 SEK / 25 tests


Estimated Shipment
03-12-2020
Add to basket

Description

Description

CD300e is a member of the glycoprotein family of cell surface proteins expressed on myeloid cells. CD300e interacts with the TYRO protein tyrosine kinase-binding protein and is thought to be involved as an activating receptor. CD300e is expressed on mature myeloid dendritic cells and monocytes, and is down regulated on immature dendritic cells. Further, CD300e interacts with DAP-12, which mediates activating signals. The alloreactive response of naïve T cells has been reported to become enhanced by the activation of CD300e. Diseases associated with CD300e dysfunction include tungiasis and lice infestation.
Specifications

Specifications

CD300e (IREM-2)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q496F6
CMRF35-like molecule 2, CLM2, CD300 antigen-like family member E, CMRF35-A5, Immune receptor expressed on myeloid cells 2, IREM-2, Polymeric immunoglobulin receptor 2, PIgR-2, PIgR2, Poly-Ig receptor 2, CD300LE, CMRF35A5, IREM2, CLM-2
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
UP-H2
PE
CD300E
Liquid
CD300E
IgG1
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
342510
Videos
SDS
Documents

For Research Use Only.