missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ CD307e (FcRL5) Polyclonal Antibody

Beskrivning
In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) IRTA3, rhIRTA4, and rhIRTA5 is observed and less than 5% cross-reactivity with rhIRTA1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Fc receptor homologs (FcRH5), also known as IRTA2 proteins, are transmembrane proteins with sequence homology to classical Fc receptor families. FcRH5 function as activating or inhibitory receptors on discrete B cell populations and are involved in B-cell development and differentiation in peripheral lymphoid organs. FcRH5 expression is deregulated in multiple myeloma and Burkitt lymphoma cell lines with 1q21 abnormalities. FcRH5 may play a critical role for the pathogenesis of B-cell lymphoma.
Specifikationer
Specifikationer
| Antigen | CD307e (FcRL5) |
| Användningsområden | Flow Cytometry, Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 0.2 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 5% trehalose and No Preservative |
| Gen | FCRL5 |
| Genaccessionsnr. | Q96RD9 |
| Gene Alias | BXMAS1; BXMAS1-like protein 2; CD307; CD307e; fc receptor homolog 3; Fc receptor homolog 5; Fc receptor like 5; Fc receptor-like 3; Fc receptor-like 5; Fc receptor-like protein 3; Fc receptor-like protein 5; Fcrh3; FCRH5; Fcrl3; Fcrl5; FcR-like protein 5; Immune receptor translocation-associated protein 2; immunoglobulin superfamily receptor translocation associated 2 (IRTA2); IRTA2; mBXMH2; moFcRH3; PRO820; rCG62831-like; UNQ503/PRO820 |
| Gensymboler | FCRL5 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?