Learn More
CD309 (FLK1) Monoclonal Antibody (Avas12a1), Super Bright™ 436, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
1232.00 SEK - 3290.00 SEK
Specifications
Antigen | CD309 (FLK1) |
---|---|
Clone | Avas12a1 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15923434
|
Affymetrix eBioscience
62-5821-82 |
100 μg |
3290.00 SEK
100µg |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15913434
|
Invitrogen
62-5821-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The Avas12a1 monoclonal antibody reacts with mouse Flk-1, also known as vascular endothelial growth factor receptor 2 (VEGFR2). Flk-1 is a receptor tyrosine kinase involved in vascular endothelial tissue development and is expressed on endothelial cells during embryonic stages and some endothelial tissues in the adult. Applications Reported: This Avas12a1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This Avas12a1 antibody has been tested by flow cytometric analysis of bEnd.3 cells. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information.
CD309 (FLK1) Monoclonal antibody specifically detects CD309 (FLK1) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD309 (FLK1) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P35918 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
Avas12a1 | |
Flow Cytometry | |
Super Bright 436 | |
Rat | |
Mouse | |
16542 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |