missing translation for 'onlineSavingsMsg'
Learn More

CD320 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA568014

Additional Details : Weight : 0.01000kg

Product Code. 15816433

  • 4215.00 SEK / 100µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Predicted to react with Mouse and Rat samples.

CD320 Polyclonal antibody specifically detects CD320 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

CD320
Polyclonal
Unconjugated
Cd320
425O18-1; 8D6; 8D6 antigen; 8D6A; 8D6AFDC-SM-8D6; 8D6TCBLR; Cd320; CD320 antigen; CD320 molecule; D17Ertd716e; FDC-signaling molecule 8D6; FDC-SM-8D6; NG29; putative VLDL lipoprotein receptor; RGD1305860; TCblR; transcobalamin receptor; UNQ198/PRO224; VLDL
Rabbit
affinity chromatography
RUO
362851, 51293, 54219
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q5HZW5, Q9NPF0, Q9Z1P5
Cd320
Synthesized peptide derived from human CD320.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers