Learn More
CD321 (F11R) Monoclonal Antibody (CSTEM27), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1366.00 SEK - 3200.00 SEK
Specifications
Antigen | CD321 (F11R) |
---|---|
Clone | CSTEM27 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15885333
|
Affymetrix eBioscience
12-3219-42 |
100 Tests |
3200.00 SEK
Pack of 100 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15875333
|
Affymetrix eBioscience
12-3219-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This monoclonal antibody CSTEM27 reacts with human CD321 (F11R, JAM-1, JAM-A). Applications Reported: This CSTEM27 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CSTEM27 antibody has been pre-diluted and tested by flow cytometric analysis of human iPSCs. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD321 (F11R, JAM-1, JAM-A) is a cell-surface adhesion molecule belonging to the Ig superfamily of receptors. CD321 is a 32-41 kD glycoprotein, possessing two extracellular Ig-like domains, a transmembrane region, and a short cytosolic tail. The protein assembles at the membrane as a homodimer, and can associate laterally with other membrane proteins including integrins. CD321 is expressed on a variety of cell types, including leukocytes, platelets, erythrocytes, stem cells, epithelial and endothelial cells, and plays a role in mediating cell-cell contact. CD321 can be found in close proximity to several components of epithelial tight junctions, including ZO-1 and occludin. CD321 is involved in the maintenance of epithelial and endothelial barriers, hematopoiesis, angiogenesis, and tumor proliferation, invasion, and metastasis.Specifications
CD321 (F11R) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q9Y624 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
CSTEM27 | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
50848 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |