missing translation for 'onlineSavingsMsg'
Learn More

CD326 (EpCAM), Alexa Fluor 488, clone: MH99, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-8326-42

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15520257

  • 2375.00 SEK / 100 tests


Estimated Shipment date
from Supplier 25-07-2018
Add to basket

Description

Description

Description: The monoclonal antibody MH99 recognizes human CD326 also known as EpCAM (Epithelial cell adhesion molecule). This 40kDa type I transmembrane protein is involved in cell-cell interactions in a calcium-independent manner. EpCAM is expressed primarily on the basolateral surface of most epithelia. Although normal epithelia express low levels of CD326, increased expression has been correlated with increased proliferation and progression to a mesenchymal phenotype. CD326 has also been used as a diagnostic marker on circulating metastatic carcinoma cells, while cancer cells of non-epithelial origin do not express EpCAM.Based on crossblocking studies, MH99 and 1B7 see overlapping epitopes.Applications Reported: This MH99 antibody has been reported for use in flow cytometric analysis.Applications Tested: This MH99 antibody has been pre-titrated and tested by flow cytometric analysis of the A549 cell line. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD326 (EpCAM)
Monoclonal
5μl/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P16422
Epithelial Specific Antigen, ESA, KSA, M4S1, MK-1, DIAR5, EGP-2, EGP40, KS1/4, MIC18, TROP1, TACSTD1, EPCAM, Epithelial cell adhesion molecule
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MH99
Alexa Fluor 488
EPCAM
Conjugated
EPCAM
IgG2a, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
4072
Videos

For Research Use Only.