Läs mer
Invitrogen™ CD335 (NKp46) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589869
Beskrivning
Immunogen sequence: SMFLLLKEGR SSHVQRGYGK VQAEFPLGPV TTAHRGTYRC FGSYNNHAWS FPSEPVKLLV TGDIENTSLA PEDPTFPDTW GTYLLTTETG LQKDHALWDH TAQNLLRMGL AFLVLVALVW FLVEDWLSRK RTRERASRAS TWEGRRRLNT QTL; Positive Samples: A-549, Mouse spleen, Mouse thymus, Rat spleen, Rat lung; Cellular Location: Cell membrane, Single-pass type I membrane protein.
CD335, also known as NKp46 or NCR1, is a member of the natural cytotoxicity receptor family, which includes NKp30 and NKp44. This type I transmembrane glycoprotein is expressed on NK cells and plays a crucial role in activating and expanding resting NK cells through interactions with dendritic cells. CD335 enhances the ability of NK cells to eliminate tumor and virus-infected cells, such as those infected by influenza. CD335 consists of two Ig-like domains and is related to leukocyte immunoglobulin-like receptors (LIR) and killer inhibitory receptors (KIR). It contributes to the increased efficiency of activated NK cells in mediating tumor cell lysis. Additionally, CD335 interacts with pancreatic beta cells during the development of type 1 diabetes, highlighting its involvement in both immune defense and autoimmune processes.
Specifikationer
| CD335 (NKp46) | |
| Polyclonal | |
| Unconjugated | |
| NCR1 | |
| 1-Mar; activating receptor 1; Ar1; CD335; hNKp46; Kilr1; KILR-1; LY94; Ly-94; lymphocyte antigen 94; Lymphocyte antigen 94 homolog; lymphocyte antigen 94 homolog (activating NK-receptor; lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46); mAR-1; MAR1 (mouse activating receptor 1); mNKp46; natural cytotoxicity triggering receptor 1; Natural killer cell p46-related protein; natural killer cell receptor; Ncr1; NK cell-activating receptor; NK receptor KILR-1; NKACTR; Nkp46; NK-p46; NK-p46); rAR-1; rat-activating receptor 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 117547, 17086, 9437 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.41 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O76036, Q8C567, Q9Z0H5 | |
| NCR1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 151-303 of human NCR1 (NP_0011389292). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.