Learn More
CD337 (NKp30) Monoclonal Antibody (AF29-4D12), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 25-3379-42
2363.45 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.23750kg
Description
Description: This AF29-4D12 monoclonal antibody reacts with human CD337, which is also known as NKp30. Expressed on NK cells, CD337 is the predomit member of the natural cytotoxicity receptor family that also includes NKp46 and NKp44. CD337 is involved in activating and expanding resting NK cells following interaction with dendritic cells to eliminate tumor, virus-infected, and immature dendritic cells. Several ligands for this receptor have been reported, including B7-H6. The AF29-4D12 antibody has been reported to crossreact to rhesus macaque. Applications Reported: This AF29-4D12 antibody has been reported for use in flow cytometric analysis. Applications Tested: This AF29-4D12 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.
CD337 (NKp30) Monoclonal antibody specifically detects CD337 (NKp30) in Human, Non-human Primate, Rhesus Monkey samples. It is validated for Flow Cytometry.Specifications
CD337 (NKp30) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
O14931, Q8MJ02 | |
Ncr3 | |
Affinity chromatography | |
RUO | |
259197, 715574 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
AF29-4D12 | |
PE-Cyanine7 | |
Ncr3 | |
1C7; Activating natural killer receptor p30; activating NK-A1 receptor; CD337; LY117; lymphocyte antigen 117; MALS; Natural cytotoxicity triggering receptor 3; natural killer cell p30-related protein; natural killer cell receptor; Ncr3; NK receptor 1c7; Nkp30; NK-p30; NKp30S; NKp30v1; NKp30v1ED | |
Mouse | |
100 Tests | |
Primary | |
Human, Non-human Primate, Rhesus Monkey | |
Antibody | |
IgG1 κ |
For Research Use Only.