missing translation for 'onlineSavingsMsg'
Learn More

CD357 (AITR/GITR) Mouse anti-Human, eFluor® 450, Clone: eBioAITR, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-5875-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15559776

  • 988.00 SEK / 25 tests


Estimated Shipment
13-01-2021
Add to basket

Description

Description

Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered. Glucocorticoid-induced tumor necrosis factor receptor family-related gene (GITR) is a member of the tumor necrosis factor receptor (TNFR) family that is expressed at low levels on unstimulated T cells, B cells, and macrophages. Upon activation, CD4+ and CD8+ T cells up-regulate GITR expression, whereas immunoregulatory T cells constitutively express high levels of GITR.
Specifications

Specifications

CD357 (AITR/GITR)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9Y5U5
AITR, GITR-D, TNFRSF18
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBioAITR
eFluor™ 450
TNFRSF18
Liquid
TNFRSF18
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
8784
Videos
SDS
Documents

For Research Use Only.