missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CD36 (Human) Recombinant Protein
Human CD36 full-length ORF ( NP_000063.2, 1 a.a. - 472 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000948-P02.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MGCDRNCGLIAGAVIGAVLAVFGGILMPVGDLLIQKTIKKQVVLEEGTIAFKNWVKTGTEVYRQFWIFDVQNPQEVMMNSSNIQVKQRGPYTYRVRFLAKENVTQDAEDNTVSFLQPNGAIFEPSLSVGTEADNFTVLNLAVAAASHIYQNQFVQMILNSLINKSKSSMFQVRTLRELLWGYRDPFLSLVPYPVTTTVGLFYPYNNTADGVYKVFNGKDNISKVAIIDTYKGKRNLSYWESHCDMINGTDAASFPPFVEKSQVLQFFSSDICRSIYAVFESDVNLKGIPVYRFVLPSKAFASPVENPDNYCFCTEKIISKNCTSYGVLDISKCKEGRPVYISLPHFLYASPDVSEPIDGLNPNEEEHRTYLDIEPITGFTLQFAKRLQVNLLVKPSEKIQVLKNLKRNYIVPILWLNETGTIGDEKANMFRSQVTGKINLLGLIEMILLSVGVVMFVAFMISYCACRSKTIK
Specifications
NP_000063.2 | |
CD36 molecule (thrombospondin receptor) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD36 | |
GST |
948 | |
Wheat germ expression system | |
25 μg | |
CHDS7, FAT, GP3B, GP4, GPIV, PASIV, SCARB3 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |