missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD368 (Clec6) Monoclonal Antibody (9B9), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1436.00 SEK - 3585.00 SEK
Specifications
Antigen | CD368 (Clec6) |
---|---|
Clone | 9B9 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15550077
|
Affymetrix eBioscience
50-9113-41 |
25 Tests |
1436.00 SEK
25 tests |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15560077
|
Affymetrix eBioscience
50-9113-42 |
100 Tests |
3585.00 SEK
100 tests |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.Specifications
CD368 (Clec6) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
CD368; Clec4d; CLEC6; CLEC-6; CLECSF8; CLECSF8Mpcl; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 8; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 8; C-type lectin domain family 4 member D; C-type lectin domain family 4, member d; C-type lectin receptor; C-type lectin superfamily member 8; C-type lectin, superfamily member 8; C-type lectin-like receptor 6; macrophage C-type lectin; macrophage-restricted C-type lectin; Mcl; MGC40078; Mpcl; superfamilymember 8 | |
CLEC4D | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CLEC4D |
9B9 | |
Flow Cytometry | |
eFluor 660 | |
Mouse | |
Human | |
Q8WXI8 | |
338339 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.