missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ CD37 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585590
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Keep as concentrated solution. Positive Control: human CD37-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It may play a role in T-cell-B-cell interactions. Alternate splicing results in multiple transcript variants encoding different isoforms.
Specifikationer
| CD37 | |
| Polyclonal | |
| Unconjugated | |
| CD37 | |
| CD37; CD37 antigen; CD37 molecule; cell differentiation antigen 37; GP52-40; Leukocyte antigen CD37; leukocyte surface antigen CD37; tetraspanin-26; Tspan26; Tspan-26 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 951 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P11049 | |
| CD37 | |
| Recombinant protein encompassing a sequence within the center region of human CD37. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering