Learn More
CD370 (Clec9A) Monoclonal Antibody (9A11), APC-eFluor™ 780, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
4395.00 SEK
Specifications
Antigen | CD370 (Clec9A) |
---|---|
Clone | 9A11 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15792476
|
Affymetrix eBioscience
47-3709-42 |
100 Tests |
4395.00 SEK
Pack of 100 |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This monoclonal antibody 9A11 reacts with human CD370, also known as C-type lectin family member 9A (Clec9A) and DNGR1. This type II transmembrane glycoprotein is present at the cell surface in the form of a homodimer. CD370 is a cellular receptor expressed by myeloid cells that recognizes F-actin, a dead cell-associated antigen that is exposed in necrotic cells. Upon ligation, CD370 recruits Syk kinase to the ITAM-like motif in its intracellular tail and facilitates endocytosis of antigenic material that is subsequently processed and cross-presented to CD8+ T cells. Human CD370 is expressed by CD141+ (BDCA3+) dendritic cells and, to a lesser degree, by CD14+ CD16- monocytes. Unlike in mice, human plasmacytoid dendritic cells do not express CD370. This 9A11 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. APC-eFluor™ 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochrome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.
CLEC9A is a group V C-type lectin-like receptor (CTLR) that functions as an activation receptor and is expressed on myeloid lineage cells.Specifications
CD370 (Clec9A) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
Q6UXN8 | |
CLEC9A | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CLEC9A |
9A11 | |
Flow Cytometry | |
APC-eFluor 780 | |
Mouse | |
Human | |
283420 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only