missing translation for 'onlineSavingsMsg'
Learn More

CD38, Alexa Fluor 700, clone: HIT2, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 56-0389-42

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15510357

  • 2575.00 SEK / 100 tests


EU Stock 2
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The HIT2 monoclonal antibody reacts with the human CD38 molecule, an approximately 45kDa type II transmembrane protein. CD38 is expressed by thymocytes, peripheral B cells including plasma cells, activated T cells, and monocytes. CD38 is a counter-receptor for CD31 and has both cyclase and hydrolase enzymatic activity.Applications Reported: This HIT2 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HIT2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Alexa Fluor® 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD38
HIT2
Alexa Fluor 700
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.5μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
ADP-Ribosyl Cyclase, Cyclic ADP-Ribose Hydrolase
IgG1
RUO
Primary
Human
Videos