missing translation for 'onlineSavingsMsg'
Learn More

CD38 Mouse anti-Human, PE, Clone: HB7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0388-42

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15566766

  • 1922.00 SEK / 100 tests


In stock, ready to ship: 4
Add to basket

Description

Description

Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. CD38 (NAD+ glycohydrolase) is a type II transmembrane glycoprotein able to induce activation, proliferation and differentiation of mature lymphocytes and mediate apoptosis of myeloid and lymphoid progenitor cells. CD38 functions as a multi-catalytic ectoenzyme serving as ADP-ribosyl cyclase, cyclic ADP-ribose hydrolase and possibly NAD+ glycohydrolase or as a cell surface receptor. Antibodies to CD38 are useful in subtyping of lymphomas and leukemias, detection of plasma cells (i.e. identification of myelomas), and as a marker for activated B and T cells. CD38 participates in cell adhesion, signal transduction and calcium signaling. Further, CD38 is expressed at high levels in the pancreas, liver, kidney, maligt lymphoma and neuroblastoma. Diseases associated with CD38 dysfunction include chronic lymphocytic leukemia and Richter’s Syndrome.
Specifications

Specifications

CD38
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P28907
T10, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, CD38 antigen p45, NAD+ nucleosidase, cADPr hydrolase 1, cyclic ADP-ribose hydrolase 2
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HB7
PE
CD38
Liquid
CD38
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
952
Videos
SDS
Documents

For Research Use Only.