missing translation for 'onlineSavingsMsg'
Learn More

CD3D & CD3E Recombinant Mouse Protein, hIgG1-Fc & hIgG1-Fc.His tag

CD3D & CD3E Recombinant Mouse Protein, hIgG1-Fc & hIgG1-Fc.His tag

Brand:  Invitrogen CT025M2303H25

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15461114

  • 2545.00 SEK / 25µg


Estimated Shipment date
from Supplier 03-11-2017
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

CD3 Antigen delta and CD3 Antigen epsilon Heterodimer (CD3D & CD3E), recombinant mouse protein is supplied as a lyophilized powder.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 105) of mouse CD3D (P04235) fused to the C-terminal flag-tagged Fc region of human IgG1 at the C-terminus, constructed the plasmid 1; a DNA sequence encoding the extracellular domain (Met 1-Asp 108) of mouse CD3E (P22646) fused to the C-terminal His-tagged Fc region of human IgG1 at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the mouse CD3D/CD3E heterodimer was purified.

Activity: This recombinant protein has not been tested.

Formulation: 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by SINO Biological.

Specifications

Specifications

Protein assays and analysis
Lyophilized
76kDa
7.4 ±0.5
≥85% by SDS-PAGE
–20°C
5 Pack(s)
CD3D & CD3E Recombinant Mouse Protein
CD3D & CD3E
CD proteins
CD3D & CD3E Recombinant Mouse Protein
DYKDDDDK, His Tag, IgG1 Fc Tag, hIgG1-Fc & hIgG1-Fc.His Tag
Lyophilized
12500, 12501
CD3D & CD3E Recombinant Mouse Protein
PAGE
5 x 5μg
For research use only. Not for use in diagnostic procedures
None
5 Vials of lyophilized protein
Assay standard
Mammalian
Mouse