missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
CD4 Antibody (C4/206) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47674-0.1MG
4743.00 SEK valid until 2025-12-16
BÄSTA PRIS på kampanj! Använd kampanjkod: "24090" för att få ditt kampanjpris.
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for ELISA.
Specifikationer
| CD4 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready | |
| CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Recombinant human CD4 protein | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a protein of 55kDa, identified as CD4. CD4 is a membrane glycoprotein of T lymphocytes that interacts with major histocompatibility complex class II antigens and is also a receptor for the human immunodeficiency virus. This protein is expressed not only in T lymphocytes, but also in B cells, macrophages, and granulocytes. It is also expressed in specific regions of the brain. The protein functions to initiate or augment the early phase of T-cell activation, and may function as an important mediator of indirect neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcript variants encoding different isoforms have been identified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, CyTOF | |
| C4/206 | |
| Unconjugated | |
| PBS with No Preservative | |
| Mouse | |
| 55 kDa | |
| 0.1 mg | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Immunology, Innate Immunity, Regulatory Immunology, Stem Cell Markers | |
| 920 | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering