Läs mer
IBA Lifesciences CD4 Fab-Strep
Handla allt IBA Lifesciences Produkter
Art:
Human
Mouse
Förpackningsstorlek:
50µg
Beskrivning
- CD4 Fab-Strep is a recombinantly expressed low affinity Fab fragment fused to a Twin-Strep-tag™
- Due to its low affinity for the target, an increase in avidity is required to ensure sufficient binding to the cell surface
- Combining Fab-Streps with a Strep-Tactin™ backbone leads to the formation of complexes that stably bind to cells
- Fab-Streps were developed for Traceless Affinity Cell Selection (Fab-TACS™), which is based on our Strep-tag™ technology
Specifikationer
Specifikationer
| Form | Lyophilized |
| För användning med (applikation) | Fluorescent Cell Staining & Sorting, Magnetic or Affinity Chromatographic Cell Isolation |
| Art | Mouse |
| Tillräckligt för | Murine CD4 |