missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Rat anti-Mouse, BUV395, Clone: GK1.5, BD Horizon™
Rat Monoclonal Antibody
Brand: BD Biosciences 565974
Additional Details : Weight : 0.01000kg
Description
CD4 Monoclonal antibody specifically detects CD4 in Mouse samples. It is validated for Flow cytometry.Storage Buffer: Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Specifications
CD4 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
25 μg | |
Cell Biology | |
Mouse | |
Purified | |
QC Testing: Mouse |
Flow Cytometry | |
GK1.5 | |
Brilliant Ultraviolet 395 | |
Cd4; CD4 antigen; L3T4; Ly-4; T-cell surface antigen T4/Leu-3 | |
Mouse CTL clone V4 | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity and unconjugated antibody and free BD Horizon™ BUV395 were removed. chromatography. The antibody was conjugated with BD Horizon™ BUV395 under optimum conditions | |
IgG2b κ |
For Reseach Use Only.