missing translation for 'onlineSavingsMsg'
Learn More

CD41a Mouse anti-Human, APC, Clone: HIP8, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0419-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15577926

  • 816.00 SEK / 25 tests


In stock, ready to ship: 1
Add to basket

Description

Description

CD41 (platelet glycoprotein IIb, ITGA2B) is composed of two subunits -120 kDa a, alpha and 23 kDa b, beta- that interact with CD61 in the presence of calcium to form a functional adhesive protein receptor. CD41 is also involved in blood coagulation by mediating platelet aggregation. Upon blood vessel damage, CD41 binds to a variety of proteins including von Willebrand factor, fibrinogen, fibronectin and vitronectin. CD41 is mainly expressed on megakaryocyte-platelet lineage, but generally belongs to the antigens that are expressed during the early stages of hematopoietic differentiation. Diseases associated with CD41 dysfunction include Glansmann Thrombasthenia, and Platelet type-16 bleeding disorder.
Specifications

Specifications

CD41a
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P08514
BDPLT2, CD41, CD41B, GP2B, GPIIb, GT, GTA, HPA3, GPalpha IIb, integrin alpha-IIb, platelet fibrinogen receptor, alpha subunit, platelet membrane glycoprotein IIb, platelet-specific antigen BAK, ITGA2B
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HIP8
APC
ITGA2B
Liquid
ITGA2B
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3674
Videos
SDS
Documents

For Research Use Only.