Learn More
CD42a Monoclonal Antibody (GR-P), eFluor™ 450, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1224.00 SEK - 2505.00 SEK
Specifications
Antigen | CD42a |
---|---|
Clone | GR-P |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15559676
|
Affymetrix eBioscience
48-0428-41 |
25 Tests |
1224.00 SEK
25 tests |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15569676
|
Affymetrix eBioscience
48-0428-42 |
100 Tests |
2505.00 SEK
100 tests |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This GR-P monoclonal antibody reacts with human CD42a, which is also known as glycoprotein IX (GPIX). CD42a is a 22-kDa transmembrane protein that is constitutively expressed on platelets and megakaryocytes. CD42a associates with CD42b, CD42c, and CD42d to form the GPIb-V-IX complex, which binds the von Willebrand factor, an endothelial adhesion protein, to mediate platelet adhesion and activation. This binding results in platelet aggregation at sites of vascular injury. This receptor complex has also been demonstrated to bind thrombin, factors XI and XII, P-selectin, and Mac-1. Finally, a point mutation within the leucine-rich motif of CD42a has been linked to Bernard-Soulier syndrome. Applications Reported: Anti-Human CD42a eFluor™ 450 has been reported for use in flow cytometric analysis. Applications Tested: The GR-P antibody has been pre-titrated and tested by flow cytometric analysis of normal human platelets. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor™ 450 is an alternative to Pacific Blue™. eFluor™ 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.
CD42a Monoclonal antibody specifically detects CD42a in Human samples. It is validated for Flow Cytometry.Specifications
CD42a | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
CD42a; Glycoprotein 9; glycoprotein 9 (platelet); glycoprotein IX (platelet); glycoprotein IX platelet; GP9; GPIX; GP-IX; platelet glycoprotein IX; Platelet GPIX | |
GP9 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
GP9 |
GR-P | |
Flow Cytometry | |
eFluor 450 | |
Mouse | |
Human | |
P14770 | |
2815 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.