missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD42c Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CD42c |
---|---|
Concentration | 180 μg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896635
|
Proteintech
55161-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16886635
|
Proteintech
55161-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The CD42c antibody from Proteintech is a rabbit polyclonal antibody to a peptide of human CD42c. This antibody recognizes human antigen. The CD42c antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
CD42c | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
2812 |
180 μg/mL | |
Polyclonal | |
Liquid | |
Human | |
Primary |