missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ CD42d Polyclonal Antibody
Beskrivning
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human GPVI is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells.
Specifikationer
Specifikationer
| Antigen | CD42d |
| Användningsområden | Flow Cytometry, Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 0.2 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 5% trehalose and No Preservative |
| Gen | Gp5 |
| Genaccessionsnr. | P40197 |
| Gene Alias | CD42d; Glycoprotein 5; glycoprotein 5 (platelet); glycoprotein 5, platelet; glycoprotein V (platelet); glycoprotein V platelet; GP5; GPV; Platelet glycoprotein 5; platelet glycoprotein V; Platelete glycoprotein 5; Platelets GP V; PLGPV |
| Gensymboler | Gp5 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?