Learn More
CD43/T-Cell Marker Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry and flow cytometry applications with Epredia™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-146-R7
Additional Details : Weight : 0.01200kg
Description
CD43 (leukosialin, sialophorin, or leukocyte sialoglycoprotein) is a cell surface glycoprotein which is expressed on all thymocytes and T-cells. CD43 is involved in activation of T cells, B cells, NK cells, and monocytesHost Species: Mouse
Clone: DF-T1
Isotype: IgG1
Species Reactivity: Human. Does not react with rat. Others not tested
Epitope: Not determined
Immunogen: Myeloblastic KG1 cells
Molecular Weight: 95/115/135 kDa (depending upon extent of glycosylation)
Positive Control: Tonsil or lymph node
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Flow Cytometry
Specifications
CD43/T-Cell Marker Ab-1 | |
Monoclonal | |
Unconjugated | |
Myeloblastic KG1 cells | |
IVD | |
Primary | |
Prediluted |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
DF-T1 | |
Mouse | |
7 mL | |
T-Cell | |
Human | |
IgG1 |