missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Lab Vision™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible reults in immunohistochemistry and flow cytometry applications with Thermo Scientific™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody.

Brand:  Thermo Scientific™ MS-146-P

Additional Details : Weight : 0.00250kg

 View more versions of this product

Product Code. 12669886

  • 2600.00 SEK / 1mL


Estimated Shipment
04-09-2020
Add to basket

Description

Description

CD43 (leukosialin, sialophorin, or leukocyte sialoglycoprotein) is a cell surface glycoprotein which is expressed on all thymocytes and T-cells. CD43 is involved in activation of T cells, B cells, NK cells, and monocytes

Host Species: Mouse

Clone: DF-T1

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not tested

Epitope: Not determined

Immunogen: Myeloblastic KG1 cells

Molecular Weight: 95/115/135 kDa (depending upon extent of glycosylation)

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry
Specifications

Specifications

CD43/T-Cell Marker Ab-1
Monoclonal
200µg/mL
Purified Ab with BSA and Azide
Murine
IgG1
IVD
Primary
Monoclonal
Flow Cytometry, Immunohistochemistry (Paraffin)
DF-T1
Unconjugated
Concentrated
Myeloblastic KG1 cells
1mL
T-Cell
Primary
Human
Videos
Documents