missing translation for 'onlineSavingsMsg'
Learn More

CD43/T-Cell Marker Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry and flow cytometry applications with Epredia™ CD43/T-Cell Marker Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-146-P0

Additional Details : Weight : 0.00170kg

 View more versions of this product

Product Code. 12679886

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

CD43 (leukosialin, sialophorin, or leukocyte sialoglycoprotein) is a cell surface glycoprotein which is expressed on all thymocytes and T-cells. CD43 is involved in activation of T cells, B cells, NK cells, and monocytes

Host Species: Mouse

Clone: DF-T1

Isotype: IgG1

Species Reactivity: Human. Does not react with rat. Others not tested

Epitope: Not determined

Immunogen: Myeloblastic KG1 cells

Molecular Weight: 95/115/135 kDa (depending upon extent of glycosylation)

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Flow Cytometry
Specifications

Specifications

CD43/T-Cell Marker Ab-1
Monoclonal
200µg/mL
Mouse
100 μL
T-Cell
Human
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin)
DF-T1
Unconjugated
Myeloblastic KG1 cells
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions