missing translation for 'onlineSavingsMsg'
Läs mer

STEMCELL Technologies CD437

Produktkod. 17160112
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
25 mg
Förpackningsstorlek:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
17160112 5 mg 5mg
17170112 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17160112 Leverantör STEMCELL Technologies Leverantörsnummer 72722
 Begränsad artikel
Beräknad leverans: 25-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Retinoid pathway activator; Activates retinoic acid receptor (RAR)

  • CD437 is the prototypical adamantyl arotinoid of the retinoid-related molecule family that acts as a selective agonist of retinoic acid receptor (RAR)γ (Kd = 6.5 μM, 2.5 μM, and 77 nM for RARα, β, and γ, respectively; Bernard et al.; Pérez-Rodríguez et al.)
  • Increases speed and number of pre-induced pluripotent stem (iPS) cell colonies generated from mouse embryonic fibroblasts (MEFs) transfected with Oct4, Sox2, c-Myc, and Klf4 (Wang et al.)
  • Induces cell cycle arrest and apoptosis in a variety of cancer cells (Fontana and Rishi; Jin et al.; Li et al.; Valli et al.)
  • Decreases mRNA expression of squamous differentiation markers cytokeratin 1, involucrin, and SPR1 in the human head and neck squamous cell carcinoma cell line, UMSCC22B (Sun et al.)

Specifikationer

CAS 125316-60-1
Synonym AHPN
Kvantitet 5 mg
Molekylformel C27H26O3
Renhet ≥95%
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.