missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44 Monoclonal Antibody (KM201), APC, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen A14713
Additional Details : Weight : 0.01000kg
Description
CD44 Monoclonal antibody specifically detects CD44 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD44 | |
Monoclonal | |
0.1 mg/mL | |
PBS with <0.1% sodium azide | |
P15379 | |
CD44 | |
(C57 BL/6 x DBA/2)F1 mouse bone marrow derived stromal cells clone BM52. | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
KM201 | |
APC | |
CD44 | |
AU023126; AW121933; AW146109; Bovine CD44; CD44; CD44 antigen; CD44 antigen-like protein; CD44 molecule; CD44 molecule (Indian blood group); CD44 protein; CD44 variant; CD44A; CD44-like protein; CDW44; cell surface glycoprotein CD44; Cell surface glycoprotein CD44 (hyaluronate binding protein); cell-surface glycoprotein; chondroitin sulfate proteoglycan 8; CSPG8; ECMRIII; ECMR-III; Epican; extracellular matrix receptor III; extracellular matrix receptor-III; GP90 lymphocyte homing/adhesion receptor; HCELL; hematopoietic cell E- and L-selectin ligand; heparan sulfate proteoglycan; HERMES; Hermes antigen; homing function and Indian blood group system; HUTCH-I; hyaluronan receptor; Hyaluronate receptor; IN; LHR; Ly-24; lymphocyte antigen 24; lymphocyte surface antigen precursor CD44; MC56; MDU2; MDU3; METAA; MIC4; PGP I; Pgp1; PGP-1; PGPI; PGP-I; Phagocytic glycoprotein 1; phagocytic glycoprotein I; putative Bos taurus CD44; RHAMM; sCD 44; sCD44; soluble CD 44; soluble CD44 | |
Rat | |
purified | |
RUO | |
12505 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |