missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 / LCA Mouse, Clone: PTPRC/1460, enQuireBio™
Mouse Monoclonal Antibody
2050.00 SEK - 5730.00 SEK
Specifications
Antigen | CD45 / LCA |
---|---|
Clone | PTPRC/1460 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960748
|
EnquireBio
5788-MSM20-P1 |
100 μg |
5730.00 SEK
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950748
|
EnquireBio
5788-MSM20-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
CD45 / LCA | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
5788 | |
Human B-lymphocytes | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
180-220kDa |
PTPRC/1460 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_002829 | |
PTPRC | |
IgG1 κ | |
Protein A/G Purified | |
Cell surface and cytoplasmic | |
Ramos, U-698, or GA-10 cells. Tonsil |