Läs mer
Invitrogen™ CD45 Monoclonal Antibody (30-F11), Functional Grade, eBioscience™

Rat Monoclonal Antibody
2765.00 SEK
Specifikationer
| Antigen | CD45 |
|---|---|
| Klona | 30-F11 |
| Koncentration | 1 mg/mL |
| Innehåll och lagring | 4°C |
| Användningsområden | Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
15218457
![]() |
Invitrogen™
16-0451-85 |
500 μg |
2765.00 SEK
500µg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
Beskrivning
Description: The 30-F11 monoclonal antibody reacts with all isoforms of mouse CD45, also known as Leukocyte Common Antigen (LCA). CD45 is expressed by all hematopoietic cells excluding mature erythrocytes and platelets. The cytoplasmic portion of CD45 has tyrosine phosphatase enzymatic activity and plays an important role in activation of lymphocytes. Applications Reported: The 30-F11 antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining. 30-F11 has also been reported in complement-dependent cytotoxicity. (Please use Functional Grade purified 30-F11 in functional assays). Applications Tested: This 30-F11 antibody has been tested by flow cytometric analysis of mouse bone marrow cells and splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase (PTP) ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 is absent on non-hematopoietic cell lines, normal and malignant, non-hematopoietic tissues. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing, differ in their extracellular domains but share identical transmembrane and cytoplasmic domains. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45 has been shown to be an essential regulator of T- and B-cell antigen receptor signaling and suppresses JAK kinases to regulate cytokine receptor signaling. CD45 is also important in promoting cell survival by modulating integrin-mediated signal transduction pathway, DNA fragmentation during apoptosis and inhibition or upregulation of various immunological functions.Specifikationer
| CD45 | |
| 1 mg/mL | |
| Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation | |
| Functional Grade | |
| Rat | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| P06800 | |
| 19264 | |
| Primary | |
| Affinity chromatography |
| 30-F11 | |
| 4°C | |
| Monoclonal | |
| Liquid | |
| IgG2b κ | |
| Mouse | |
| PTPRC | |
| B220; cd45; CD45 antigen; CD45 antigen isoform 1 precursor; CD45 antigen isoform 2 precursor; CD45 antigen isoform 3 precursor; CD45 antigen isoform 4 precursor; CD45 antigen isoform 5 precursor; CD45 antigen isoform 6 precursor; CD45R; GP180; Lca; L-CA; leucocyte common antigen; leukocyte common antigen; leukocyte common antigen A; leukocyte common antigen B; leukocyte common antigen, CD45; loc; LOW QUALITY PROTEIN: receptor-type tyrosine-protein phosphatase C; LY5; Ly-5; lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase lambda; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; Protein tyrosine phosphatase, receptor-type, c polypeptide; protein tyrosine phosphatase; alternatively spliced; Ptprc; Receptor-type tyrosine-protein phosphatase C; RT7; T200; T200 glycoprotein; T200 leukocyte common antigen; T220 and B220 | |
| PTPRC | |
| Antibody |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.