Läs mer
CD45 Mouse anti-Human, Clone: 2D1, BD™
Beskrivning
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow cytometry.
Specifikationer
Specifikationer
| Antigen | CD45 |
| Användningsområden | Flow Cytometry |
| Klassificering | Monoclonal |
| Klona | 2D1 |
| Koncentration | 25μg/mL |
| Konjugera | Unconjugated |
| Beskrivning | Vol. Per test: 20μL |
| Formulering | Phosphate buffered saline with gelatin and 0.1% sodium azide. |
| Värd art | Mouse |
| Kvantitet | 100 Tests |
| Visa mer |
For Reseach Use Only.