Learn More
CD45R (B220) Monoclonal Antibody (RA3-6B2), PE-eFluor™ 610, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
2740.00 SEK
Specifications
Antigen | CD45R (B220) |
---|---|
Clone | RA3-6B2 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15570437
|
Affymetrix eBioscience
61-0452-82 |
100 μg |
2740.00 SEK
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The RA3-6B2 monoclonal antibody reacts with exon A-restricted isoform of mouse CD45, a 220 kDa surface molecule. CD45R/B220 epitope is mainly expressed by the B cell lineage from early Pro-B to mature B cells. However, some activated T cells, lymphokine activated killer cells (LAK), NK cell progenitors in the bone marrow, and T cells of the lpr/lpr mutant mouse also express this antigen. Applications Reported: This RA3-6B2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RA3-6B2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PE-eFluor™ 610 can be excited with laser lines from 488-561 nm and emits at 607 nm. We recommend using a 610/20 band pass filter (equivalent to PE-Texas Red™). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat.
CD45R (B220) Monoclonal antibody specifically detects CD45R (B220) in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
CD45R (B220) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P06800, P08575 | |
PTPRC | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
PTPRC |
RA3-6B2 | |
Flow Cytometry | |
PE-eFluor 610 | |
Rat | |
Human, Mouse | |
19264, 5788 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.