missing translation for 'onlineSavingsMsg'
Learn More

CD45R (B220) Rat anti-Human, Mouse, PE-eFluor® 610, Clone: RA3-6B2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 61-0452-80

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15560437

  • 928.00 SEK / 25µg


Estimated Shipment
11-01-2021
Add to basket

Description

Description

00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. CD45R is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. CD45R contains an extracellular domain, a single transmembrane segment and two tandem intracytoplasmic catalytic domains. It is specifically expressed in hematopoietic cells and has been shown to be an essential regulator of T- and B-cell antigen receptor signaling.
Specifications

Specifications

CD45R (B220)
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P06800, P08575
B220, CD45R, Cd45, Ly-5, Lyt-4, T200, Cluster designation 45R
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Flow Cytometry
RA3-6B2
PE-eFluor™ 610
PTPRC
Liquid
PTPRC
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
19264, 5788
Videos
SDS
Documents

For Research Use Only.