Learn More
CD45RB Monoclonal Antibody (PD7/26), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1642.00 SEK - 4340.00 SEK
Specifications
Antigen | CD45RB |
---|---|
Clone | PD7/26 |
Concentration | 0.5 mg/mL |
Classification | Monoclonal |
Conjugate | Alexa Fluor 488 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15520277
|
Affymetrix eBioscience
53-9458-82 |
100 μg |
4340.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15510277
|
Affymetrix eBioscience
53-9458-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal antibody PD7/26 recognizes CD45RB, which is an isoform of the Leukocyte Common Antigen (LCA) also known as CD45. CD45RB is a protein tyrosine phosphatase receptor that is present on most hematopoietic cells and absent on non-hematopoietic cells. Expression of CD45RB is more restricted than CD45 and can only be found on T cell subsets, B, NK, and myeloid cells. Regulatory T cells (Tregs) have been show to express low levels of CD45RB, which may correlate with increased migration to sites of infection. In contrast, high CD45RB expression results in IL-2 and IFN gamma production instead of IL-4 in vitro. Applications Reported: This PD7/26 antibody has been reported for use in flow cytometric analysis, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry. Applications Tested: This PD7/26 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue using low pH antigen retrieval and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD45RB Monoclonal antibody specifically detects CD45RB in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
CD45RB | |
0.5 mg/mL | |
Alexa Fluor 488 | |
Mouse | |
Human | |
5788 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
PD7/26 | |
Monoclonal | |
Liquid | |
RUO | |
P08575 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.