Learn More
CD47 Monoclonal Antibody (miap301), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 16-0471-050
Additional Details : Weight : 0.06000kg
Description
The MAb miap301 reacts to CD47 also known as integrin-associated protein (IAP), and neurophilin. CD47 is a glycosylated five transmembrane protein with a small cytoplasmic domain. CD47 is involved in adhesion through interactions with SIRP (signal regulator protein) and is non-covalently associated with ?3 integrins CD51/CD61, CD41/CD61. It is also known to be the receptor for thrombospondin. T cell expression of CD47 can mediate activation or apoptosis (in the presence of high levels of thrombospondin). Expression is found in the majority of hematopoietic cells including T and B cells, monocytes, platelets and erythrocytes (as part of the Rh complex). Expression is also found in non-hematopoietic cells. No crossreactivity is seen against human CD47.
CD47 Monoclonal antibody specifically detects CD47 in Mouse samples. It is validated for Flow Cytometry, Functional Assay, NeutralizationSpecifications
CD47 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
Q61735 | |
Rat | |
50 mg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry, Functional Assay, Neutralization | |
miap301 | |
Functional Grade | |
CD47 | |
9130415E20Rik; AA407862; AI848868; antigen identified by monoclonal antibody 1D8; antigenic surface determinant protein OA3; AW108519; B430305P08Rik; Cd47; CD47 antigen; CD47 antigen (Rh-related antigen, integrin-associated signal transducer); CD47 antigen/integrin-associated protein; CD47 glycoprotein; Cd47 molecule; CD47/IAP; IAP; integrin associated protein; integrin-associated protein; integrin-associated signal transducer; Itgp; Leukocyte surface antigen CD47; MER6; OA3; Protein MER6; Rh-related antigen; sCD47; soluble CD 47; soluble CD47 | |
Affinity chromatography | |
RUO | |
16423 | |
4° C | |
Liquid |
For Research Use Only.