missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGA2 Monoclonal Antibody (DX5), PE, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen A14741
Additional Details : Weight : 0.01000kg
Description
ITGA2 Monoclonal antibody specifically detects ITGA2 in Mouse samples. It is validated for Flow Cytometry.Specifications
ITGA2 | |
Monoclonal | |
0.1 mg/mL | |
PBS with <0.1% sodium azide | |
Q62469 | |
Itga2 | |
RIL-2-propagated NK1.1 cells from C57BL/6 mice. | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgM κ |
Flow Cytometry | |
DX5 | |
PE | |
Itga2 | |
alpha 2 subunit of VLA-2 receptor; BDPLT9; BR; CD49 antigen-like family member B; CD49B; Collagen receptor; DX5; GPIa; HPA-5; human platelet alloantigen system 5; integrin alpha 2; integrin alpha-2; integrin subunit alpha 2; integrin, alpha 2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); Itga2; platelet antigen Br; platelet glycoprotein GPIa; platelet membrane glycoprotein Ia; very late activation protein 2 receptor, alpha-2 subunit; VLA-2; VLA2 receptor; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha; VLAA2 | |
Rat | |
purified | |
RUO | |
16398 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |