missing translation for 'onlineSavingsMsg'
Learn More

CD49d (Integrin alpha 4) Mouse anti-Human, Clone: 9F10, eBioscience™

Mouse Monoclonal Antibody

458.00 SEK - 920.00 SEK

Specifications

Antigen CD49d (Integrin alpha 4)
Clone 9F10
Target Species Human
Conjugate Unconjugated
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15207017
View Documents
Affymetrix eBioscience
14-0499-80
25 μg
458.00 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 09-09-2021
Please to purchase this item. Need a web account? Register with us today!
15217017
View Documents
Affymetrix eBioscience
14-0499-82
100 μg
920.00 SEK
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 09-09-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The 9F10 monoclonal antibody reacts with human CD49d, the 150 kDa integrin alpha4 subunit. The complex of CD49d non-covalently associated with integrin beta1 (CD29), also known as VLA-4, is a receptor for fibronectin and VCAM-1 (CD106). This complex is expressed by thymocytes, peripheral lymphocytes, monocytes and eosinophils. CD49d also associates with integrin beta7 and binds to the Mucosal Addressin Cell-Adhesion Molecule-1 (MadCAM-1). Applications Reported: The 9F10 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining. 9F10 has also been reported in in vitro functional assays. (Please use Functional Grade purified 9F10, cat. 16-0499, in functional assays.). Applications Tested: The 9F10 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

The integrin alpha 4 (also known as CD49d and ITGA4) belongs to the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha and beta chains. Alpha4 chain associates with either beta1 or beta7 chain. It has been demonstrated that the putative ligand-binding sites of both integrin alpha4beta1 and alpha4beta7 is located on the alpha4 chain. These ligands included Madcam, VCAM, and fibronectin. Madcam is known as the principal ligand for integrin alpha4beta7. Recently it was also demonstrated that HIV-1 envelope can mimic Madcam by binding to and signaling though integrin alpha4beta7, the gut mucosal homing receptor for peripheral T cells.
Specifications

Specifications

CD49d (Integrin alpha 4)
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C
9F10
Unconjugated
Flow Cytometry, Functional Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Primary
Monoclonal
PBS with 0.09% sodium azide; pH 7.2
RUO
SDS
Product Certifications

For Research Use Only.