missing translation for 'onlineSavingsMsg'
Learn More

CD49d (Integrin alpha 4) Mouse anti-Human, PE, Clone: 9F10, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0499-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15566786

  • 796.00 SEK / 25 tests


Estimated Shipment
08-12-2020
Add to basket

Description

Description

The integrin alpha 4 (also known as CD49d and ITGA4) belongs to the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha and beta chains. Alpha4 chain associates with either beta1 or beta7 chain. It has been demonstrated that the putative ligand-binding sites of both integrin alpha4beta1 and alpha4beta7 is located on the alpha4 chain. These ligands included Madcam, VCAM, and fibronectin. Madcam is known as the principal ligand for integrin alpha4beta7. Recently it was also demonstrated that HIV-1 envelope can mimic Madcam by binding to and signaling though integrin alpha4beta7, the gut mucosal homing receptor for peripheral T cells.
Specifications

Specifications

CD49d (Integrin alpha 4)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P13612
269C wild type, CD49 antigen-like family member D, VLA-4 subunit alpha, antigen CD49D, alpha-4 subunit of VLA-4 receptor, ITGA4, ITGA-IV
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
9F10
PE
ITGA4
Liquid
ITGA4
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3676
Videos
SDS
Documents

For Research Use Only.