Learn More
CD49d (Integrin alpha 4) Monoclonal Antibody (9F10), Super Bright™ 702, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1460.00 SEK - 3365.00 SEK
Specifications
Antigen | CD49d (Integrin alpha 4) |
---|---|
Clone | 9F10 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933734
|
Affymetrix eBioscience
67-0499-42 |
100 Tests |
3365.00 SEK
Pack of 100 |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923734
|
Affymetrix eBioscience
67-0499-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This CF1D12 monoclonal antibody reacts with human CD52, a cell surface glycoprotein expressed in the immune and reproductive systems. CF1D12 is thought to recognize the carbohydrate region of CD52, not the protein core of the molecule. Tissue-specific modifications of the glycan structure may alter epitope recognition in certain applications. For example, this CF1D12 clone has been reported to recognize human sperm glycoforms of CD52 by western blot, but not by flow cytometry. The immunogen used to generate this antibody was human CD52. Applications Reported: This CF1D12 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CF1D12 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 702 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 702 nm. We recommend using a 710/50 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No.
CD49d (Integrin alpha 4) Monoclonal antibody specifically detects CD49d (Integrin alpha 4) in Human samples. It is validated for Flow Cytometry.Specifications
CD49d (Integrin alpha 4) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P13612 | |
ITGA4 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ITGA4 |
9F10 | |
Flow Cytometry | |
Super Bright 702 | |
Mouse | |
Human | |
3676 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |