Learn More
CD5 Monoclonal Antibody (UCHT2), Alexa Fluor™ 700, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1510.00 SEK - 3155.00 SEK
Specifications
Antigen | CD5 |
---|---|
Clone | UCHT2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16626424
|
Affymetrix eBioscience
56-0059-42 |
100 Tests |
3155.00 SEK
100 tests |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16616424
|
Affymetrix eBioscience
56-0059-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The UCHT2 monoclonal antibody reacts with human CD5, a 67 kDa protein expressed by a majority of thymocytes and mature T cells and a subset of B cells. Signaling through the CD5 molecule activates T cells and binding of CD5 to its ligand on B cells, CD72, and plays an important role in T-B interaction and proliferation. The monoclonal antibody UCHT2 recognizes primate CD5. Applications Reported: This UCHT2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This UCHT2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Alexa Fluor 700 emits at 723 nm and can be excited with the red laser (633-647 nm). Most instruments will require a 685 LP mirror and 710/20 band pass filter. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser.
CD5 Monoclonal antibody specifically detects CD5 in Human samples. It is validated for Flow Cytometry.Specifications
CD5 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
921 | |
IgG1 κ | |
Affinity chromatography | |
Cd5 |
UCHT2 | |
Flow Cytometry | |
Alexa Fluor 700 | |
Mouse | |
Human | |
P06127 | |
Cd5 | |
Primary | |
Antibody |