Learn More
CD5 Mouse anti-Human, Unconjugated, Clone: 4D12C10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67278-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16839815
  • 3313.85 SEK / 150µL
Estimated Shipment: 05-10-2022
to see stock.
Description
Description
The CD5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD5. This antibody recognizes human antigen. The CD5 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Specifications
CD5
Monoclonal
1000 μg/mL
Mouse
Primary
Human
ELISA, Immunofluorescence, Immunohistochemistry
4D12C10
Unconjugated
150 μL
921
IgG2b