missing translation for 'onlineSavingsMsg'
Learn More

CD5 Rat anti-Canine, PerCP-eFluor™ 710, Clone: YKIX322.3, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-5050-41

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15579256

  • 922.00 SEK / 25 tests


Estimated Shipment
11-12-2020
Add to basket

Description

Description

Our testing indicates that PerCP-eFluor® 710 conjugated antibodies are stable when stained samples are exposed to freshly prepared 2% formaldehyde overnight at 4°C, but please evaluate for alternative fixation protocols. Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered. CD5 is a 67 kDa human T-lymphocyte single-chain transmembrane glycoprotein. CD5 is present on all mature T-lymphocytes, on most of thymocytes and on many T-cell leukemias and lymphomas. CD5 also reacts with a subpopulation of activated B-cells and may act as a receptor in regulating T-cell proliferation. CD5 is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Diseases associated with CD5 dysfunction include thymus cancer and Richter’s Syndrome.
Specifications

Specifications

CD5
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Liquid
CD28
IgG2a, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
403646
Flow Cytometry
YKIX322.3
PerCP-eFluor™ 710
CD28
CD5 antigen p56-62, T-cell surface glycoprotein CD5, lymphocyte antigen T1/Leu-1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Canine
Videos
SDS
Documents

For Research Use Only.