missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD62E/CD62P Monoclonal Antibody (1.2B6), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen A15437
Additional Details : Weight : 0.00280kg
Description
Clone 1.2B6 recognizes a common epitope shared by human CD62P and CD62E.
CD62E/CD62P Monoclonal antibody specifically detects CD62E/CD62P in Human, Porcine samples. It is validated for Flow Cytometry.Specifications
CD62E/CD62P | |
Monoclonal | |
0.1 mg/mL | |
PBS with <0.1% sodium azide | |
P16109, P16581, P98110 | |
SELE, SELP | |
Human CD62E | |
100 μg | |
Primary | |
Human, Porcine | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
1.2B6 | |
Biotin | |
SELP | |
CD62; CD62 antigen-like family member E; CD62 antigen-like family member P; CD62E; CD62P; CD62P; CD62 homolog; ELAM; ELAM1; ELAM-1; endothelial adhesion molecule 1; endothelial leukocyte adhesion molecule 1; ESEL; E-selectin; GMP140; GMP-140; GMRP; Granule membrane protein 140; granule membrane protein 140kDa; granulocyte membrane protein; GRMP; LECAM2; LECAM-2; LECAM3; LECAM-3; leukocyte endothelial cell adhesion molecule 2; leukocyte-endothelial cell adhesion molecule 2; Leukocyte-endothelial cell adhesion molecule 3; PADGEM; Platelet activation dependent granule-external membrane protein; platelet alpha-granule membrane protein; PSEL; P-selectin; P-selectin precursor; RP1-117P20.2; SELE; selectin E; selectin E (endothelial adhesion molecule 1); selectin P; selectin P (granule membrane protein 140kDa, antigen CD62); SELP | |
Mouse | |
purified | |
RUO | |
397150, 397508, 6401, 6403 | |
4° C | |
Liquid |