missing translation for 'onlineSavingsMsg'
Learn More

CD68 Rat anti-Mouse, eFluor® 660, Clone: FA-11, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-0681-80

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15569886

  • 1190.00 SEK / 25µg


Estimated Shipment
04-01-2021
Add to basket

Description

Description

Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. This FA-11 antibody has been tested using frozen mouse tissue and can be used at less than or equal to 10 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 µm post-manufacturing filtered. CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predomitly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human maligcies, including several Acute Myeloid Leukemia studies.
Specifications

Specifications

CD68
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P31996
Cd68
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12514
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
FA-11
eFluor™ 660
Cd68
Liquid
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Videos
SDS
Documents

For Research Use Only.