Learn More
CD69 Monoclonal Antibody (CH/4), PE-Cyanine5.5, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MHCD6918
Additional Details : Weight : 0.50000kg
Description
The R-phycoerythrin (PE)-Cy5.5® tandem conjugate permits simultaneous multicolor labeling and detection of multiple targets with excitation by a single excitation source-the 488 nm line of the argon-ion laser.
CD69 Monoclonal antibody specifically detects CD69 in Human samples. It is validated for Flow Cytometry.Specifications
CD69 | |
Monoclonal | |
PE-Cyanine5.5 | |
CD69 | |
5830438K24Rik; Activation inducer molecule; activation inducer molecule (AIM/CD69); AI452015; AIM; BL-AC/P26; CD69; CD69 antigen; CD69 antigen (p60, early T-cell activation antigen); CD69 molecule; CLEC2C; C-type lectin domain family 2 member C; C-type lectin domain family 2, member C; EA1; early activation antigen CD69; early lymphocyte activation antigen; early T-cell activation antigen p60; GP32/28; leukocyte surface antigen Leu-23; MLR-3; VEA; Very Early Activation Antigen | |
Mouse | |
purified | |
ASR | |
969 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
CH/4 | |
PBS with BSA, sucrose and 0.1% sodium azide | |
Q07108 | |
CD69 | |
Human CD69 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |