Läs mer
CD7 Antibody (15), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89748PE
4993.75 SEK valid until 2025-12-16
BÄSTA PRIS på kampanj! Använd kampanjkod: "24090" för att få ditt kampanjpris.
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
CD7 Monoclonal antibody specifically detects CD7 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Specifikationer
| CD7 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse (Negative), Rat (Negative) | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 15 | |
| ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA Capture | |
| CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CD7 (Accession#: NP_006128.1; Met 1-Pro 180). | |
| 0.1 mL | |
| Cytokine Research, Signal Transduction | |
| 924 | |
| Store at 4C in the dark. | |
| IgG |