missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD73 Monoclonal Antibody (AD2), Super Bright™ 600, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1422.00 SEK - 3250.00 SEK
Specifications
Antigen | CD73 |
---|---|
Clone | AD2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15933524
|
Affymetrix eBioscience
63-0739-42 |
100 Tests |
3250.00 SEK
Pack of 100 |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923524
|
Affymetrix eBioscience
63-0739-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This AD2 monoclonal antibody reacts with human CD73, a 5'-ectonucleotidase that converts 5'-adenosine monophosphate to adenosine. CD73 is expressed on the surface of endothelial cells, as well as B and T cells, including some CD4+Foxp3+ regulatory T cells. Adenosine production by these cells has been linked to the inhibition of CD4 T cell effector functions such as proliferation and cytokine secretion.
CD73 (Ecto-5-prime-nucleotidase, 5-prime-ribonucleotide phosphohydrolase) catalyzes the conversion at neutral pH of purine 5-prime mononucleotides to nucleosides, the preferred substrate being AMP. CD73 consists of a dimer of 2 identical 70 kDa subunits bound externally to the plasma membrane by a glycosyl phosphatidyl inositol linkage. CD73 is used as a marker of lymphocyte differentiation. Consequently, a deficiency of CD73 occurs in a variety of immunodeficiency diseases. Other forms of 5-prime nucleotidase exist in the cytoplasm and lysosomes and can be distinguished from CD73 by their substrate affinities, requirement for divalent magnesium ion, activation by ATP, and inhibition by inorganic phosphate. The CD73 gene has been localized to 6q14-q21 by immunofluorescence and a study of a panel of human x mouse hybrids that contained fragments of chromosome 6 as translocations. Defects in the CD73 gene can lead to the calcification of joints and arteries, and intestinal tuberculosis. Two transcript variants encoding different isoforms of CD73 have been found.Specifications
CD73 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P21589 | |
NT5E | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
NT5E |
AD2 | |
Flow Cytometry | |
Super Bright 600 | |
Mouse | |
Human | |
4907 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |